COM448640143 Combi-Blocks, Inc.

tert-Butyldimethyl(2-(thiophen-2-yl)ethoxy)silane

  • MDL number MFCD11501907
  • Cas Number : 160744-11-6
  • Molecular Weight : 242.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-4934

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-4934
Availability details for
Quantity
  • Availability
  • Add to Cart