COM448640169 Combi-Blocks, Inc.

1,6-Dibromoperfluorohexane

  • MDL number MFCD00153110
  • Cas Number : 918-22-9
  • Molecular Weight : 459.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-4972

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-4972
Availability details for
Quantity
  • Availability
  • Add to Cart