COM448640276 Combi-Blocks, Inc.

5-Bromo-1-benzofuran-2-carbaldehyde

  • MDL number MFCD06659072
  • Cas Number : 23145-16-6
  • Molecular Weight : 225

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5109

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5109
Availability details for
Quantity
  • Availability
  • Add to Cart