COM448640418 Combi-Blocks, Inc.

N-Alpha-phthalyl-l-glutamine

  • MDL number MFCD00190993
  • Cas Number : 3343-29-1
  • Molecular Weight : 276.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5286

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5286
Availability details for
Quantity
  • Availability
  • Add to Cart