COM448640550 Combi-Blocks, Inc.

Imidazolyl-4-ethanol

  • MDL number MFCD00195671
  • Cas Number : 872-82-2
  • Molecular Weight : 112.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5433

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5433
Availability details for
Quantity
  • Availability
  • Add to Cart