COM448640566 Combi-Blocks, Inc.

N-Boc-beta-alanine-beta-methyl-n-carboxyanhydride

  • MDL number MFCD03839892
  • Cas Number : 357610-31-2
  • Molecular Weight : 229.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5449

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5449
Availability details for
Quantity
  • Availability
  • Add to Cart