COM448640703 Combi-Blocks, Inc.

tert-Butyl 1-benzhydryl-3-azetidinylcarbamate

  • MDL number MFCD06409217
  • Cas Number : 91189-18-3
  • Molecular Weight : 338.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5600

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5600
Availability details for
Quantity
  • Availability
  • Add to Cart