COM448640710 Combi-Blocks, Inc.

N-(4-Amino-3-methoxyphenyl)methanesulfonamide hcl

  • MDL number MFCD03931062
  • Cas Number : 83209-83-0
  • Molecular Weight : 252.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5609

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5609
Availability details for
Quantity
  • Availability
  • Add to Cart