COM448640757 Combi-Blocks, Inc.

Sodium 4-bromophenylmethanesulfonate

  • MDL number MFCD11113780
  • Cas Number : 64755-47-1
  • Molecular Weight : 273.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5665

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5665
Availability details for
Quantity
  • Availability
  • Add to Cart