COM448640784 Combi-Blocks, Inc.

N-Boc-beta-alanine-alpha-methyl-n-carboxyanhydride

  • MDL number MFCD03839893
  • Cas Number : 357610-32-3
  • Molecular Weight : 229.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5696

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5696
Availability details for
Quantity
  • Availability
  • Add to Cart