COM448640825 Combi-Blocks, Inc.

N-Hexyl-2-pyrrolidinone

  • MDL number MFCD01310443
  • Cas Number : 4838-65-7
  • Molecular Weight : 169.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5748

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5748
Availability details for
Quantity
  • Availability
  • Add to Cart