COM448640842 Combi-Blocks, Inc.

tert-Butoxycarbonylamino-(1,4-dioxa-spiro[4.5]dec-8-yl)-acetic acid

  • MDL number MFCD04115606
  • Cas Number : 876761-75-0
  • Molecular Weight : 315.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5771

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5771
Availability details for
Quantity
  • Availability
  • Add to Cart