COM448640877 Combi-Blocks, Inc.

4-N-Propylpyridine

  • MDL number MFCD00014639
  • Cas Number : 1122-81-2
  • Molecular Weight : 121.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5818

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5818
Availability details for
Quantity
  • Availability
  • Add to Cart