COM448640948 Combi-Blocks, Inc.

Methyl d-4-chlorophenylglycinate

  • MDL number MFCD07784110
  • Cas Number : 43189-43-1
  • Molecular Weight : 199.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5920

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-5920
Availability details for
Quantity
  • Availability
  • Add to Cart