COM448641133 Combi-Blocks, Inc.

Trans-(4-aminomethylcyclohexyl)acetic acid hcl

  • MDL number MFCD17926317
  • Cas Number : 1228880-64-5
  • Molecular Weight : 207.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-6145

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-6145
Availability details for
Quantity
  • Availability
  • Add to Cart