COM448641535 Combi-Blocks, Inc.

tert-Butyl (s)-1-(4-(ethoxycarbonyl)oxazol-5-yl)ethylcarbamate

  • MDL number MFCD09025770
  • Cas Number : 672310-07-5
  • Molecular Weight : 284.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-6613

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-6613
Availability details for
Quantity
  • Availability
  • Add to Cart