COM448641901 Combi-Blocks, Inc.

Tri(2-methoxycarbonylethyl)phosphine

  • MDL number MFCD07369992
  • Cas Number : 29269-17-8
  • Molecular Weight : 292.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7028

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7028
Availability details for
Quantity
  • Availability
  • Add to Cart