COM448641904 Combi-Blocks, Inc.

tert-Butyl 6-carboxy-4-oxospiro[chroman-2,4'-piperidine]-1'-carboxylate

  • MDL number MFCD11501220
  • Cas Number : 921760-85-2
  • Molecular Weight : 361.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7035

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7035
Availability details for
Quantity
  • Availability
  • Add to Cart