COM448642224 Combi-Blocks, Inc.

2-(3-Benzoylphenyl)propionitrile

  • MDL number MFCD00044435
  • Cas Number : 42872-30-0
  • Molecular Weight : 235.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7418

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7418
Availability details for
Quantity
  • Availability
  • Add to Cart