COM448642306 Combi-Blocks, Inc.

Nomifensine

  • MDL number MFCD00242716
  • Cas Number : 24526-64-5
  • Molecular Weight : 238.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7525

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7525
Availability details for
Quantity
  • Availability
  • Add to Cart