COM448642431 Combi-Blocks, Inc.

tert-Butyl 4-(2-((tert-butoxycarbonyl)amino)ethyl)benzoate

  • MDL number MFCD20261460
  • Cas Number : 1334499-61-4
  • Molecular Weight : 321.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7684

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7684
Availability details for
Quantity
  • Availability
  • Add to Cart