COM448642589 Combi-Blocks, Inc.

tert-Butyl 4-(2-(p-tolylsulfonyl)hydrazono)piperidine-1-carboxylate

  • MDL number MFCD11100927
  • Cas Number : 1046478-89-0
  • Molecular Weight : 367.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7899

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7899
Availability details for
Quantity
  • Availability
  • Add to Cart