COM448642593 Combi-Blocks, Inc.

Trans-4-amino-1-benzyl-3-hydroxymethyl piperidine

  • MDL number MFCD03839871
  • Cas Number : 1177198-30-9
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7903

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7903
Availability details for
Quantity
  • Availability
  • Add to Cart