COM448642595 Combi-Blocks, Inc.

N,N-Dimethyl-4-oxo-3,4-dihydroquinazoline-7-carboxamide

  • MDL number MFCD11501912
  • Cas Number : 1092460-54-2
  • Molecular Weight : 217.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7906

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7906
Availability details for
Quantity
  • Availability
  • Add to Cart