COM448642652 Combi-Blocks, Inc.

N-(4-Carbethoxyphenyl)-n-(3-cyanophenyl)amine

  • MDL number MFCD04115332
  • Cas Number : 458550-46-4
  • Molecular Weight : 266.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7979

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7979
Availability details for
Quantity
  • Availability
  • Add to Cart