COM448642661 Combi-Blocks, Inc.

tert-Butyl 3-(4-methoxyphenyl)piperazine-1-carboxylate

  • MDL number MFCD06660434
  • Cas Number : 886768-17-8
  • Molecular Weight : 292.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7991

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-7991
Availability details for
Quantity
  • Availability
  • Add to Cart