COM448642739 Combi-Blocks, Inc.

3-(6-Bromopyridin-2-yl)benzoic acid

  • MDL number MFCD05864809
  • Cas Number : 914349-44-3
  • Molecular Weight : 278.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8092

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8092
Availability details for
Quantity
  • Availability
  • Add to Cart