COM448642759 Combi-Blocks, Inc.

N-Boc-2,6-dimethyl-l-tyrosine

  • MDL number MFCD01860643
  • Cas Number : 99953-00-1
  • Molecular Weight : 309.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8114

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8114
Availability details for
Quantity
  • Availability
  • Add to Cart