COM448642796 Combi-Blocks, Inc.

N-(4'-Carboxylic)benzoyl-4-piperidone

  • MDL number MFCD00798179
  • Cas Number : 672309-93-2
  • Molecular Weight : 247.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8153

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8153
Availability details for
Quantity
  • Availability
  • Add to Cart