COM448642840 Combi-Blocks, Inc.

N-(5-Acetyl-2-hydroxy-phenyl)-methanesulfonamide

  • MDL number MFCD06796623
  • Cas Number : 14347-18-3
  • Molecular Weight : 229.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8206

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8206
Availability details for
Quantity
  • Availability
  • Add to Cart