COM448642904 Combi-Blocks, Inc.

Methanedisulfonic acid

  • MDL number MFCD00041888
  • Cas Number : 503-40-2
  • Molecular Weight : 176.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8279

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8279
Availability details for
Quantity
  • Availability
  • Add to Cart