COM448643117 Combi-Blocks, Inc.

N-(3-Aminomethyl-phenyl)-2-(4-hydroxy-phenyl)-acetamide

  • MDL number MFCD07369839
  • Cas Number : 886363-58-2
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8529

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8529
Availability details for
Quantity
  • Availability
  • Add to Cart