COM448643211 Combi-Blocks, Inc.

N-Boc-3,5-difluoro-d-tyrosine

  • MDL number MFCD08689604
  • Cas Number : 1213198-51-6
  • Molecular Weight : 317.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8651

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8651
Availability details for
Quantity
  • Availability
  • Add to Cart