COM448643431 Combi-Blocks, Inc.

2-Amino-2-methylpropanenitrile

  • MDL number MFCD00043024
  • Cas Number : 19355-69-2
  • Molecular Weight : 84.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8910

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8910
Availability details for
Quantity
  • Availability
  • Add to Cart