COM448643437 Combi-Blocks, Inc.

N-(3-Hydroxypyridin-2-yl)pivalamide

  • MDL number MFCD07781130
  • Cas Number : 177744-83-1
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8920

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8920
Availability details for
Quantity
  • Availability
  • Add to Cart