COM448643443 Combi-Blocks, Inc.

tert-Butyl 3-(4-bromophenyl)-2-((methylamino)methyl)propanoate, HCl

  • MDL number MFCD08061156
  • Cas Number : 1159822-24-8
  • Molecular Weight : 364.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8927
Availability details for
Quantity
  • Availability
  • Add to Cart