COM448643456 Combi-Blocks, Inc.

N-Boc-2-amino-5-bromobenzaldehyde

  • MDL number MFCD11617307
  • Cas Number : 199273-16-0
  • Molecular Weight : 300.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8946

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8946
Availability details for
Quantity
  • Availability
  • Add to Cart