COM448643459 Combi-Blocks, Inc.

Methyl-(1-phenyl-3-pyrrolidin-1-yl-propyl)-amine

  • MDL number MFCD03426435
  • Cas Number : 672310-28-0
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8949

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8949
Availability details for
Quantity
  • Availability
  • Add to Cart