COM448643473 Combi-Blocks, Inc.

Trans-(4-methyl-cyclohexyl)-acetic acid

  • MDL number MFCD06796533
  • Cas Number : 7132-93-6
  • Molecular Weight : 156.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8969

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8969
Availability details for
Quantity
  • Availability
  • Add to Cart