COM448643475 Combi-Blocks, Inc.

Methyl n-acetyl-3-(2,5-difluorophenyl)-d-alaninate

  • MDL number MFCD08436171
  • Cas Number : 1017294-05-1
  • Molecular Weight : 257.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8972

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8972
Availability details for
Quantity
  • Availability
  • Add to Cart