COM448643476 Combi-Blocks, Inc.

N-(3-Cyano-phenyl)-2-(4-hydroxy-phenyl)-acetamide

  • MDL number MFCD06796570
  • Cas Number : 886362-51-2
  • Molecular Weight : 252.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8973

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8973
Availability details for
Quantity
  • Availability
  • Add to Cart