COM448643478 Combi-Blocks, Inc.

N-(2-Amino-ethyl)-2-(benzyl-phenyl-amino)-acetamide maleate

  • MDL number MFCD06796537
  • Cas Number : 1185241-40-0
  • Molecular Weight : 399.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8975

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-8975
Availability details for
Quantity
  • Availability
  • Add to Cart