COM448643531 Combi-Blocks, Inc.

N-4-Boc-2-piperazinecarboxylic acid tert-butyl ester

  • MDL number MFCD03001707
  • Cas Number : 438631-75-5
  • Molecular Weight : 286.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9043

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9043
Availability details for
Quantity
  • Availability
  • Add to Cart