COM448643543 Combi-Blocks, Inc.

N-Boc-2-(4-trifluoromethyl-phenyl)-dl-glycine

  • MDL number MFCD07388841
  • Cas Number : 847147-40-4
  • Molecular Weight : 319.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9057

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9057
Availability details for
Quantity
  • Availability
  • Add to Cart