COM448643574 Combi-Blocks, Inc.

9-Anthracenecarbonyl chloride

  • MDL number MFCD00045385
  • Cas Number : 16331-52-5
  • Molecular Weight : 240.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9092

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9092
Availability details for
Quantity
  • Availability
  • Add to Cart