COM448643581 Combi-Blocks, Inc.

Pyridine-3,5-dicarboxylic acid monoethyl ester

  • MDL number MFCD01928035
  • Cas Number : 84254-37-5
  • Molecular Weight : 195.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9099

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9099
Availability details for
Quantity
  • Availability
  • Add to Cart