COM448643606 Combi-Blocks, Inc.

Methyl cbz-amino(diethoxyphosphoryl)acetate

  • MDL number MFCD09878835
  • Cas Number : 114684-69-4
  • Molecular Weight : 359.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9130

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9130
Availability details for
Quantity
  • Availability
  • Add to Cart