COM448643608 Combi-Blocks, Inc.

1-Boc-5-cyano-3-hydroxymethylindole

  • MDL number MFCD05864719
  • Cas Number : 914349-11-4
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9132

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9132
Availability details for
Quantity
  • Availability
  • Add to Cart