COM448643617 Combi-Blocks, Inc.

Methyl-(1-pyrrolidin-1-ylmethyl-cyclohexyl)-amine

  • MDL number MFCD03426434
  • Cas Number : 675602-55-8
  • Molecular Weight : 196.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9142

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9142
Availability details for
Quantity
  • Availability
  • Add to Cart