COM448643654 Combi-Blocks, Inc.

1-(4-Chloro-2-nitrophenyl)piperazine

  • MDL number MFCD00640777
  • Cas Number : 405910-34-1
  • Molecular Weight : 241.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9191

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-9191
Availability details for
Quantity
  • Availability
  • Add to Cart